Assay Details
| Gene Symbol: Arhgef25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166413.1 | NP_001159885.1 | 10 - 11 | 1052 | 69 | |
| NM_028027.3 | NP_082303.2 | 10 - 11 | 1114 | 69 | ||
| GenBank mRNA | AF487515.1 | - | 10 - 11 | 1080 | 69 | |
| AK010452.1 | - | 10 - 11 | 1080 | 69 | ||
| AK158684.1 | - | 10 - 11 | 1052 | 69 | ||
| BC023367.1 | - | 1 - 2 | 91 | 69 | ||
| BC041113.1 | - | 8 - 9 | 1428 | 69 | ||
| BC054833.1 | - | 2 - 3 | 336 | 6439728 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef25
Genomic Map