Assay Details
| Gene Symbol: Arhgef25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166413.1 | NP_001159885.1 | 12 - 13 | 1183 | 82 | |
| NM_028027.3 | NP_082303.2 | 12 - 13 | 1245 | 82 | ||
| GenBank mRNA | AF487515.1 | - | 12 - 13 | 1211 | 82 | |
| AK010452.1 | - | 12 - 13 | 1211 | 82 | ||
| AK158684.1 | - | 12 - 13 | 1183 | 82 | ||
| BC023367.1 | - | 3 - 4 | 222 | 82 | ||
| BC041113.1 | - | 10 - 11 | 1559 | 82 | ||
| BC054833.1 | - | 4 - 5 | 467 | 6439728 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef25
Genomic Map