Assay Details
| Gene Symbol: Arhgef25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166413.1 | NP_001159885.1 | 14 - 15 | 1570 | 65 | |
| NM_028027.3 | NP_082303.2 | 14 - 15 | 1632 | 65 | ||
| GenBank mRNA | AF487515.1 | - | 14 - 15 | 1598 | 65 | |
| AK010452.1 | - | 14 - 15 | 1598 | 65 | ||
| AK158684.1 | - | 14 - 15 | 1570 | 65 | ||
| BC023367.1 | - | 5 - 6 | 609 | 65 | ||
| BC041113.1 | - | 12 - 13 | 1946 | 65 | ||
More Information
Set Membership: |
|

Arhgef25
Genomic Map