Assay Details
| Gene Symbol: Cnot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028043.2 | NP_082319.1 | 1 - 2 | 535 | 123 | |
| GenBank mRNA | AK010503.1 | - | 1 - 2 | 547 | 123 | |
| AK144692.1 | - | 1 - 2 | 550 | 123 | ||
| AK162118.1 | - | 840 | 123 | |||
| AK167440.1 | - | 1 - 2 | 526 | 123 | ||
| BC049091.1 | - | 1 - 2 | 535 | 6401756 | 123 | |
| BC050812.1 | - | 1 - 2 | 515 | 6773595 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot11
Genomic Map