Assay Details
| Gene Symbol: Iqce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028833.3 | NP_083109.2 | 16 - 17 | 1623 | 135 | |
| GenBank mRNA | AK006472.2 | - | 15 - 16 | 1364 | 135 | |
| BC059223.1 | - | 16 - 17 | 1623 | 6417270 | 135 | |
More Information
Set Membership: |
|

Iqce
Genomic Map