Assay Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029320.3 | NP_083596.2 | 12 - 13 | 2367 | 109 | |
| GenBank mRNA | AK006060.1 | - | 3 - 4 | 341 | 109 | |
| AK017117.1 | - | 3 - 4 | 108 | |||
| AK045133.1 | - | 12 - 13 | 1864 | 109 | ||
| AK149007.1 | - | 7 - 8 | 920 | 109 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK006060.1 transcripts.
Set Membership: |
|

Pibf1
Genomic Map