Assay Details
| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029585.3 | NP_083861.1 | 1 - 2 | 102 | 103 | |
| GenBank mRNA | AK011666.1 | - | 1 - 2 | 128 | 103 | |
| BC031930.1 | - | 1 - 2 | 102 | 4018106 | 103 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK011666.1,BC031930.1,NM_029585.3 transcripts.
Set Membership: |
|

Det1
Genomic Map