Assay Details
| Gene Symbol: Smpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164609.1 | NP_001158081.1 | 5 - 6 | 469 | 153 | |
| NM_001164610.1 | NP_001158082.1 | 5 - 6 | 469 | 153 | ||
| NM_001164611.1 | NP_001158083.1 | 5 - 6 | 469 | 153 | ||
| NM_029945.3 | NP_084221.2 | 5 - 6 | 469 | 153 | ||
| GenBank mRNA | AK019466.2 | - | 5 - 6 | 470 | 153 | |
| AK031029.1 | - | 5 - 6 | 463 | 153 | ||
| AK031685.1 | - | 5 - 6 | 468 | 153 | ||
| AK035892.1 | - | 5 - 6 | 461 | 153 | ||
| AK036055.1 | - | 5 - 6 | 461 | 153 | ||
| AK036380.1 | - | 5 - 6 | 465 | 153 | ||
| AK129354.1 | - | 5 - 6 | 446 | 153 | ||
| AK138720.1 | - | 5 - 6 | 443 | 153 | ||
| AK164234.1 | - | 5 - 6 | 461 | 153 | ||
| BC026767.1 | - | 5 - 6 | 442 | 5137008 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpd4
Genomic Map