Assay Details
| Gene Symbol: Lpxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134152.3 | NP_598913.1 | 5 - 6 | 548 | 94 | |
| GenBank mRNA | AB053936.2 | - | 5 - 6 | 600 | 94 | |
| AB071194.1 | - | 5 - 6 | 573 | 94 | ||
| AK077085.1 | - | 5 - 6 | 584 | 94 | ||
| BC026563.1 | - | 5 - 6 | 514 | 5065405 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lpxn
Genomic Map