Assay Details
| Gene Symbol: Aldh16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145954.1 | NP_666066.1 | 3 - 4 | 375 | 78 | |
| GenBank mRNA | AK030673.1 | - | 3 - 4 | 368 | 78 | |
| AK144967.1 | - | 374 | 78 | |||
| AK144995.1 | - | 3 - 4 | 382 | 78 | ||
| AK151691.1 | - | 3 - 4 | 375 | 78 | ||
| AK161198.1 | - | 3 - 4 | 413 | 78 | ||
| AK164374.1 | - | 3 - 4 | 421 | 78 | ||
| AK169346.1 | - | 727 | 78 | |||
| AK170276.1 | - | 3 - 4 | 414 | 78 | ||
| AK220200.1 | - | 3 - 4 | 351 | 78 | ||
| BC013548.1 | - | 3 - 4 | 375 | 4222058 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh16a1
Genomic Map