Assay Details
| Gene Symbol: Aldh16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145954.1 | NP_666066.1 | 7 - 8 | 967 | 116 | |
| GenBank mRNA | AK030673.1 | - | 7 - 8 | 960 | 116 | |
| AK144967.1 | - | 966 | 116 | |||
| AK144995.1 | - | 7 - 8 | 974 | 116 | ||
| AK151691.1 | - | 7 - 8 | 967 | 116 | ||
| AK161198.1 | - | 7 - 8 | 1005 | 116 | ||
| AK164374.1 | - | 7 - 8 | 1013 | 116 | ||
| AK170276.1 | - | 7 - 8 | 1006 | 116 | ||
| AK220200.1 | - | 7 - 8 | 943 | 116 | ||
| BC013548.1 | - | 7 - 8 | 967 | 4222058 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh16a1
Genomic Map