Assay Details
| Gene Symbol: Aldh16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145954.1 | NP_666066.1 | 8 - 9 | 1155 | 64 | |
| GenBank mRNA | AK030673.1 | - | 8 - 9 | 1148 | 64 | |
| AK144967.1 | - | 1154 | 64 | |||
| AK144995.1 | - | 8 - 9 | 1162 | 64 | ||
| AK151691.1 | - | 8 - 9 | 1155 | 64 | ||
| AK161198.1 | - | 8 - 9 | 1193 | 64 | ||
| AK164374.1 | - | 8 - 9 | 1201 | 64 | ||
| AK169346.1 | - | 1506 | 64 | |||
| AK170276.1 | - | 8 - 9 | 1194 | 64 | ||
| AK220200.1 | - | 8 - 9 | 1131 | 64 | ||
| BC013548.1 | - | 8 - 9 | 1155 | 4222058 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh16a1
Genomic Map