Assay Details
| Gene Symbol: Ift52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172150.4 | NP_742162.2 | 9 - 10 | 842 | 84 | |
| GenBank mRNA | AK088467.1 | - | 9 - 10 | 843 | 84 | |
| AK144188.1 | - | 9 - 10 | 942 | 84 | ||
| AK167194.1 | - | 9 - 10 | 858 | 84 | ||
| AK178241.1 | - | 3 - 4 | 209 | 84 | ||
| BC037708.1 | - | 9 - 10 | 811 | 4222723 | 84 | |
More Information
Set Membership: |
|

Ift52
Genomic Map