Assay Details
| Gene Symbol: Ift52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172150.4 | NP_742162.2 | 10 - 11 | 991 | 106 | |
| GenBank mRNA | AK088467.1 | - | 10 - 11 | 992 | 106 | |
| AK144188.1 | - | 10 - 11 | 1091 | 106 | ||
| AK167194.1 | - | 10 - 11 | 1007 | 106 | ||
| AK178241.1 | - | 4 - 5 | 358 | 106 | ||
| BC037708.1 | - | 10 - 11 | 960 | 4222723 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift52
Genomic Map