Assay Details
| Gene Symbol: Foxred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291448.1 | NP_001278377.1 | 4 - 5 | 706 | 67 | |
| NM_001291449.1 | NP_001278378.1 | 4 - 5 | 711 | 67 | ||
| NM_172291.2 | NP_758495.1 | 4 - 5 | 706 | 67 | ||
| GenBank mRNA | AK054231.1 | - | 4 - 5 | 598 | 67 | |
| AK144759.1 | - | 4 - 5 | 632 | 67 | ||
| AK163722.1 | - | 4 - 5 | 565 | 67 | ||
| BC017604.1 | - | 4 - 5 | 664 | 67 | ||
| BC024806.1 | - | 4 - 5 | 576 | 5361053 | 67 | |
| BC028892.1 | - | 2 - 3 | 1677 | 67 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291449.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial respiratory chain complex I assembly
oxidation-reduction process
molecular_function
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
epoxyqueuosine reductase activity
mitochondrial respiratory chain complex I assembly
oxidation-reduction process
molecular_function
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
epoxyqueuosine reductase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26421276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26421275 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | BC027386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | BC028892.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | AK163722.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | BC017604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | BC024806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | BC027386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | BC028892.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | AK163722.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | BC017604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254583 | BC024806.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229793 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140331 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156874 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | BC027386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | BC028892.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | AK163722.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | BC017604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | BC024806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | BC027386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | BC028892.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | AK163722.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | BC017604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254583 | BC024806.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26421227 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156874 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06749345 | - | - |
| Whole Mouse Genome G4122A | A_52_P346803 | BC027386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P346803 | BC028892.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P346803 | AK163722.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P346803 | BC017604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P346803 | BC024806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | BC027386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | BC028892.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | AK163722.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | BC017604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | BC024806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | BC027386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | BC028892.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | AK163722.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | BC017604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254583 | BC024806.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102272 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156874 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07607198 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07607184 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07607150 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07607197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26421275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26421276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26421227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07607213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07607166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07722329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07656246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07836740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26421248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592124 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592121 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451354_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592114 | - | - |
| Mouse Genome 430 2.0 Array | 1451354_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451354_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235169_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235169_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900016708.mm.1 | NM_172291.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016708.mm.1 | TC0900002009.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016710.mm.1 | NM_172291.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016710.mm.1 | TC0900002009.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451354_PM_at | - | - |
| Mouse Expression Set 430 | 1451354_at | - | - |
| Murine Genome U74v2 Set | 103879_at | - | - |
| Murine Genome U74v2 Set | 103879_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451354_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451354_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451354_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451354_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 790771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3940309 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2650239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2940397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2650239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6520348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3940309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003940309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940397 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2659247 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2756957 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711754 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2659247 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2711754 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734017 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2756957 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003528.1_25-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0235169.2_28-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003454.1_20-S | - | - |
| mouse-6 v1.1 (Illumina) | scl36048.5_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650239 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520348 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940397 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3940309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003528.1_25-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003454.1_20-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0235169.2_28-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36048.5_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3940309 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520348 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650239 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2756957 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237151 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2832886 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2832884 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2711754 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237151 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2756957 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2832884 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2711754 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2832886 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003528.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0235169.2_28-S | - | - |

Foxred1
Genomic Map