Assay Details
| Gene Symbol: Cdc42se1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172395.3 | NP_765983.1 | 1 - 2 | 448 | 111 | |
| GenBank mRNA | AK004859.1 | - | 1 - 2 | 93 | 111 | |
| AK019052.1 | - | 1 - 2 | 90 | 111 | ||
| AK028573.1 | - | 1 - 2 | 90 | 111 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK004859.1,AK019052.1,AK028573.1,NM_172395.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342939 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512591 | AK028573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512591 | AK004859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512591 | AK019052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512591 | NM_172395.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512591 | AK028573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512591 | AK004859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512591 | AK019052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512591 | NM_172395.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043269 | AK028573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043269 | BC029194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043269 | BC060964.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043269 | AK004859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043269 | AK008084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043267 | AK028573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043267 | BC029194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043267 | BC060964.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043267 | AK004859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043267 | AK008084.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122685 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41926 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120135 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043267 | AK028573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043267 | BC029194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043267 | BC060964.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043267 | AK004859.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043267 | AK008084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043269 | AK028573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043269 | BC029194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043269 | BC060964.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043269 | AK004859.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043269 | AK008084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512591 | AK028573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512591 | AK004859.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512591 | AK019052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512591 | NM_172395.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512591 | AK028573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512591 | AK004859.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512591 | AK019052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512591 | NM_172395.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512591 | AK028573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512591 | AK004859.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512591 | AK019052.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512591 | NM_172395.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512591 | AK028573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512591 | AK004859.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512591 | AK019052.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512591 | NM_172395.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342960 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120135 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04994561 | - | - |
| Whole Mouse Genome G4122A | A_51_P512591 | AK028573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512591 | AK004859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512591 | AK019052.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512591 | NM_172395.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512591 | AK028573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512591 | AK004859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512591 | AK019052.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512591 | NM_172395.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P340700 | AK028573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P340700 | BC029194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P340700 | AK004859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P340700 | AK008084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P340700 | NM_172395.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105007 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02411387 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342939 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02706975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02675579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02706973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02675581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02706978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02706984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02706977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02411387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02613415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02879004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02706972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02706967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02675586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02706960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10494206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494205 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428132_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428131_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494200 | - | - |
| Mouse Genome 430 2.0 Array | 1428132_at | - | - |
| Mouse Genome 430 2.0 Array | 1428131_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428131_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428132_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57912_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57912_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006991.mm.1 | NM_172395.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006991.mm.1 | TC0300000894.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006989.mm.1 | NM_172395.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006989.mm.1 | TC0300000894.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006990.mm.1 | NM_172395.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006990.mm.1 | TC0300000894.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006988.mm.1 | NM_172395.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006988.mm.1 | TC0300000894.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006994.mm.1 | NM_001038708.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006994.mm.1 | NM_172395.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006994.mm.1 | TC0300000894.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 97817_PM_at | - | - |
| Murine Genome U74v2 Set | 97817_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428132_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428131_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428132_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428131_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 358197 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 510037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3163014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259946 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3163014 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259946 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2451048 | - | - |
| mouse-6 v1.1 (Illumina) | GI_84781705-A | - | - |
| mouse-6 v1.1 (Illumina) | scl00006.1_238-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00006.1_238-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1259946 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3163014 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3163014 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259946 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00006.1_238-S | - | - |

Cdc42se1
Genomic Map