Assay Details
| Gene Symbol: Ldah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076460.1 | - | 2 - 3 | 225 | 74 | |
| AK162850.1 | - | 2 - 3 | 190 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076460.1, AK162850.1 transcripts.
Set Membership: |
|

Ldah
Genomic Map