Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172404.2 | NP_765992.2 | 9 - 10 | 1046 | 77 | |
| GenBank mRNA | AK031511.1 | - | 9 - 10 | 1059 | 77 | |
| AK042391.1 | - | 8 - 9 | 992 | 77 | ||
| AK088404.1 | - | 8 - 9 | 954 | 77 | ||
| AK163165.1 | - | 8 - 9 | 854 | 77 | ||
| AK207151.1 | - | 3 - 4 | 290 | 77 | ||
| AK218327.1 | - | 3 - 4 | 290 | 77 | ||
| BC016206.1 | - | 9 - 10 | 1148 | 4912335 | 77 | |
| BC025124.1 | - | 8 - 9 | 1179 | 4486058 | 77 | |
| BC052047.1 | - | 9 - 10 | 1046 | 5687475 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl1
Genomic Map