Assay Details
| Gene Symbol: Pus7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172437.3 | NP_766025.1 | 4 - 5 | 1318 | 81 | |
| GenBank mRNA | AK019372.1 | - | 4 - 5 | 1311 | 81 | |
| AK029027.1 | - | 4 - 5 | 1302 | 81 | ||
| BC066798.1 | - | 3 - 4 | 1271 | 30469996 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pus7l
Genomic Map