Assay Details
| Gene Symbol: Mob3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041672.1 | - | 2255 | 65 | ||
| AK164429.1 | - | 2644 | 65 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284924 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284928 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284929 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284926 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284925 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486001 | AK030481.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486001 | AK030481.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018532 | BC058238.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018532 | AK171197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018532 | BC051170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018532 | AK170394.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018532 | NM_172457.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205194 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147297 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158423 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486001 | AK030481.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486001 | AK030481.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018532 | BC058238.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018532 | AK171197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018532 | BC051170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018532 | AK170394.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018532 | NM_172457.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486001 | AK030481.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486001 | AK030481.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P8913 | AK080169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374961 | AK041672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374961 | AK164429.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135933 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08485989 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08485993 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08485990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08485992 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284929 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284925 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284928 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284924 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08954160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08454896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08233649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08954162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08413470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08804344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08509247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08509244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284889 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10370948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370947 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434388_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10370946 | - | - |
| Mouse Genome 430 2.0 Array | 1434388_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434388_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208228_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208228_at | - | - |
| Mouse Expression Set 430 | 1434388_PM_at | - | - |
| Mouse Expression Set 430 | 1434388_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434388_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434388_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 319437 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 106180273 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6130670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253324 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2557635 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253324 | - | - |
| mouse-6 v1.1 (Illumina) | scl37722.7_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130670 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106180273 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37722.7_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130670 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106180273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3008924 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2557635 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253324 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3008924 | - | - |

Mob3a
Genomic Map