Assay Details
| Gene Symbol: Mtrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028628.1 | - | 4 - 5 | 502 | 60 | |
| AK155359.1 | - | 5 - 6 | 1215 | 60 | ||
| AK163449.1 | - | 4 - 5 | 484 | 60 | ||
| BC025942.1 | - | 457 | 5132457 | 60 | ||
| BC031565.1 | - | 4 - 5 | 498 | 60 | ||
| BC066796.1 | - | 3 - 4 | 445 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtrr
Genomic Map