Assay Details
| Gene Symbol: Mtrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028628.1 | - | 7 - 8 | 1148 | 100 | |
| AK155359.1 | - | 8 - 9 | 1861 | 100 | ||
| AK162978.1 | - | 6 - 7 | 767 | 100 | ||
| AK163449.1 | - | 7 - 8 | 1130 | 100 | ||
| BC025942.1 | - | 1103 | 5132457 | 100 | ||
| BC031565.1 | - | 7 - 8 | 1144 | 100 | ||
| BC066796.1 | - | 6 - 7 | 1091 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtrr
Genomic Map