Assay Details
| Gene Symbol: Myef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162417.1 | NP_001155889.1 | 13 - 14 | 1426 | 57 | |
| NM_010852.2 | NP_034982.2 | 12 - 13 | 1375 | 57 | ||
| GenBank mRNA | AF483504.1 | - | 11 - 12 | 1101 | 57 | |
| AF483505.1 | - | 11 - 12 | 1101 | 57 | ||
| AK048404.1 | - | 13 - 14 | 1393 | 57 | ||
| AK143230.1 | - | 12 - 13 | 1311 | 57 | ||
| U13262.1 | - | 8 - 9 | 816 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myef2
Genomic Map