Assay Details
| Gene Symbol: Fkbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012056.2 | NP_036186.2 | 7 - 8 | 1374 | 77 | |
| GenBank mRNA | AF090334.1 | - | 7 - 8 | 1300 | 77 | |
| AF279263.1 | - | 7 - 8 | 1352 | 77 | ||
| AK007499.1 | - | 5 - 6 | 764 | 77 | ||
| BC026133.1 | - | 7 - 8 | 1314 | 4018235 | 77 | |
| BC043129.1 | - | 7 - 8 | 1374 | 6810029 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp9
Genomic Map