| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800012457.mm.1 |
NM_001037877.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012457.mm.1 |
NM_001037878.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012457.mm.1 |
NM_001286362.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012457.mm.1 |
NM_001286363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012457.mm.1 |
NM_025804.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012457.mm.1 |
TC0800001520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800012459.mm.1 |
NM_001037877.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012459.mm.1 |
NM_001037878.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012459.mm.1 |
NM_001286362.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012459.mm.1 |
NM_001286363.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012459.mm.1 |
NM_025804.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012459.mm.1 |
TC0800001520.mm.1 |
MTA TC |