Assay Details
| Gene Symbol: Hcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081218.1 | NP_001074687.1 | 10 - 11 | 1595 | 77 | |
| GenBank mRNA | AK007317.1 | - | 1 - 2 | 78 | 77 | |
| BC116411.1 | - | 3 - 4 | 387 | 77 | ||
| BC127176.1 | - | 3 - 4 | 387 | 77 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK007317.1 transcripts.
Set Membership: |
|

Hcfc2
Genomic Map