Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029186.2 | NP_083462.2 | 1 - 2 | 108 | 60 | |
| GenBank mRNA | AK031554.1 | - | 1 - 2 | 54 | 60 | |
| AK151797.1 | - | 1 - 2 | 83 | 60 | ||
| AK161305.1 | - | 1 - 2 | 64 | 60 | ||
| BC031169.1 | - | 1 - 2 | 53 | 60 | ||
| BC058746.1 | - | 1 - 2 | 54 | 6399588 | 60 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK031554.1,AK151797.1,AK161305.1,BC058746.1,NM_029186.2 transcripts.
Set Membership: |
|

Tmem180
Genomic Map