Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029186.2 | NP_083462.2 | 3 - 4 | 426 | 81 | |
| GenBank mRNA | AK031554.1 | - | 3 - 4 | 372 | 81 | |
| AK151797.1 | - | 3 - 4 | 401 | 81 | ||
| AK161305.1 | - | 3 - 4 | 382 | 81 | ||
| AK170247.1 | - | 373 | 81 | |||
| BC026385.1 | - | 1 - 2 | 554 | 81 | ||
| BC031169.1 | - | 3 - 4 | 371 | 81 | ||
| BC058746.1 | - | 3 - 4 | 372 | 6399588 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem180
Genomic Map