Assay Details
| Gene Symbol: Ndfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190989.1 | NP_001177918.1 | 2 - 3 | 492 | 67 | |
| NM_029561.4 | NP_083837.2 | 2 - 3 | 492 | 67 | ||
| GenBank mRNA | AF411608.1 | - | 2 - 3 | 201 | 67 | |
| AK031073.1 | - | 2 - 3 | 307 | 67 | ||
| AK049678.1 | - | 2 - 3 | 362 | 67 | ||
| AK078939.1 | - | 2 - 3 | 402 | 67 | ||
| AK084288.1 | - | 2 - 3 | 351 | 67 | ||
| AK132235.1 | - | 2 - 3 | 421 | 67 | ||
| AK150832.1 | - | 2 - 3 | 306 | 67 | ||
| BC071237.1 | - | 2 - 3 | 411 | 5003738 | 67 | |
| BC106125.1 | - | 2 - 3 | 419 | 67 | ||
| BC117075.1 | - | 2 - 3 | 267 | 40126394 | 67 | |
| BC119092.1 | - | 2 - 3 | 267 | 8733841 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndfip2
Genomic Map