Assay Details
| Gene Symbol: Ndfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190989.1 | NP_001177918.1 | 3 - 4 | 627 | 89 | |
| NM_029561.4 | NP_083837.2 | 3 - 4 | 627 | 89 | ||
| GenBank mRNA | AF411608.1 | - | 3 - 4 | 336 | 89 | |
| AK031073.1 | - | 3 - 4 | 442 | 89 | ||
| AK049678.1 | - | 3 - 4 | 497 | 89 | ||
| AK078939.1 | - | 3 - 4 | 537 | 89 | ||
| AK132235.1 | - | 3 - 4 | 556 | 89 | ||
| AK150832.1 | - | 3 - 4 | 441 | 89 | ||
| AK220371.1 | - | 1 - 2 | 77 | 89 | ||
| BC071237.1 | - | 3 - 4 | 546 | 5003738 | 89 | |
| BC106125.1 | - | 3 - 4 | 554 | 89 | ||
| BC117075.1 | - | 3 - 4 | 402 | 40126394 | 89 | |
| BC119092.1 | - | 3 - 4 | 402 | 8733841 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndfip2
Genomic Map