Assay Details
| Gene Symbol: 3110002H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029623.2 | NP_083899.1 | 2 - 3 | 252 | 104 | |
| GenBank mRNA | AK010309.1 | - | 2 - 3 | 246 | 104 | |
| AK144623.1 | - | 2 - 3 | 253 | 104 | ||
| AK154240.1 | - | 2 - 3 | 247 | 104 | ||
| AK159691.1 | - | 2 - 3 | 247 | 104 | ||
| BC012285.1 | - | 2 - 3 | 230 | 104 | ||
| BC021846.1 | - | 2 - 3 | 235 | 5027035 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

3110002H16Rik
Genomic Map