Assay Details
| Gene Symbol: 3110002H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029623.2 | NP_083899.1 | 16 - 17 | 1565 | 102 | |
| GenBank mRNA | AF003352.1 | - | 4 - 5 | 451 | 102 | |
| AK013979.1 | - | 10 - 11 | 951 | 102 | ||
| AK144623.1 | - | 16 - 17 | 1566 | 102 | ||
| AK154240.1 | - | 16 - 17 | 1560 | 102 | ||
| AK159691.1 | - | 16 - 17 | 1560 | 102 | ||
| BC012285.1 | - | 15 - 16 | 1728 | 102 | ||
| BC021846.1 | - | 16 - 17 | 1548 | 5027035 | 102 | |
| BC044861.1 | - | 1 - 2 | 3206 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

3110002H16Rik
Genomic Map