Assay Details
| Gene Symbol: Sbf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081030.2 | NP_001074499.2 | 34 - 35 | 4929 | 107 | |
| NM_001170561.1 | NP_001164032.1 | 35 - 36 | 5007 | 107 | ||
| GenBank mRNA | AK020972.1 | - | 5 - 6 | 570 | 107 | |
| AK076039.1 | - | 2 - 3 | 174 | 107 | ||
| AK129485.1 | - | 34 - 35 | 4873 | 107 | ||
| AK147332.1 | - | 34 - 35 | 4886 | 107 | ||
| BC029156.1 | - | 5 - 6 | 565 | 107 | ||
| BC098209.1 | - | 15 - 16 | 2272 | 107 | ||
| BC157935.1 | - | 34 - 35 | 4819 | 9088115 | 107 | |
| BC158013.1 | - | 35 - 36 | 4897 | 9007320 | 107 | |
| BC172094.1 | - | 34 - 35 | 4819 | 9054380 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sbf1
Genomic Map