Assay Details
| Gene Symbol: Sbf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081030.2 | NP_001074499.2 | 38 - 39 | 5573 | 67 | |
| NM_001170561.1 | NP_001164032.1 | 39 - 40 | 5651 | 67 | ||
| GenBank mRNA | AK076039.1 | - | 6 - 7 | 818 | 67 | |
| AK129485.1 | - | 38 - 39 | 5517 | 67 | ||
| AK147332.1 | - | 38 - 39 | 5530 | 67 | ||
| BC029156.1 | - | 9 - 10 | 1209 | 67 | ||
| BC098209.1 | - | 19 - 20 | 2916 | 67 | ||
| BC157935.1 | - | 38 - 39 | 5463 | 9088115 | 67 | |
| BC158013.1 | - | 39 - 40 | 5541 | 9007320 | 67 | |
| BC172094.1 | - | 38 - 39 | 5463 | 9054380 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sbf1
Genomic Map