Assay Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153502.3 | NP_705722.2 | 7 - 8 | 747 | 106 | |
| GenBank mRNA | AF492400.1 | - | 7 - 8 | 747 | 106 | |
| BC023221.1 | - | 7 - 8 | 724 | 106 | ||
| BC132370.1 | - | 7 - 8 | 757 | 40130647 | 106 | |
| BC138797.1 | - | 7 - 8 | 757 | 8861819 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd23
Genomic Map