Assay Details
| Gene Symbol: Fuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172283.3 | NP_758487.2 | 8 - 9 | 755 | 74 | |
| NR_052009.1 | 8 - 9 | 755 | 74 | |||
| GenBank mRNA | AJ297482.4 | - | 8 - 9 | 726 | 74 | |
| AJ534942.1 | - | 8 - 9 | 726 | 74 | ||
| AK161763.1 | - | 8 - 9 | 757 | 74 | ||
| AK164340.1 | - | 8 - 9 | 724 | 74 | ||
| AY223865.1 | - | 8 - 9 | 726 | 74 | ||
| BC037698.1 | - | 7 - 8 | 704 | 4190449 | 74 | |
More Information
Set Membership: |
|

Fuk
Genomic Map