Assay Details
| Gene Symbol: Dennd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172477.4 | NP_766065.1 | 11 - 12 | 2409 | 150 | |
| GenBank mRNA | AK048041.1 | - | 11 - 12 | 2394 | 150 | |
| AK076686.1 | - | 3 - 4 | 552 | 150 | ||
| AK081176.1 | - | 11 - 12 | 2410 | 150 | ||
| AK135781.1 | - | 9 - 10 | 954 | 150 | ||
| AK160784.1 | - | 11 - 12 | 2171 | 150 | ||
| AK165202.1 | - | 11 - 12 | 2264 | 150 | ||
| AK167298.1 | - | 11 - 12 | 2263 | 150 | ||
| BC056367.1 | - | 11 - 12 | 2279 | 150 | ||
| BC057310.1 | - | 11 - 12 | 2279 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd2a
Genomic Map