Assay Details
| Gene Symbol: Dennd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172477.4 | NP_766065.1 | 16 - 17 | 3033 | 80 | |
| GenBank mRNA | AK048041.1 | - | 16 - 17 | 3018 | 80 | |
| AK076686.1 | - | 8 - 9 | 1178 | 80 | ||
| AK081176.1 | - | 16 - 17 | 3034 | 80 | ||
| AK135781.1 | - | 14 - 15 | 1578 | 80 | ||
| AK160784.1 | - | 16 - 17 | 2795 | 80 | ||
| AK165202.1 | - | 16 - 17 | 2888 | 80 | ||
| AK167298.1 | - | 16 - 17 | 2887 | 80 | ||
| BC056367.1 | - | 16 - 17 | 2903 | 80 | ||
| BC057310.1 | - | 16 - 17 | 2903 | 80 | ||
| BC145599.1 | - | 15 - 16 | 2712 | 9054165 | 80 | |
| BC150799.1 | - | 15 - 16 | 2712 | 9087710 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dennd2a
Genomic Map