Assay Details
| Gene Symbol: Sepsecs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172490.3 | NP_766078.1 | 3 - 4 | 401 | 102 | |
| GenBank mRNA | AK033654.1 | - | 3 - 4 | 402 | 102 | |
| AK049338.1 | - | 408 | 102 | |||
| AK153792.1 | - | 341 | 102 | |||
| BC062133.1 | - | 3 - 4 | 391 | 6516578 | 102 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK153792.1 transcripts.
Set Membership: |
|

Sepsecs
Genomic Map