Assay Details
| Gene Symbol: Trmt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167994.1 | NP_001161466.1 | 7 - 8 | 806 | 102 | |
| NM_172540.2 | NP_766128.2 | 6 - 7 | 730 | 102 | ||
| GenBank mRNA | AK028999.1 | - | 6 - 7 | 686 | 102 | |
| AK049523.1 | - | 6 - 7 | 728 | 102 | ||
| AK129030.1 | - | 7 - 8 | 749 | 102 | ||
| AK137595.1 | - | 692 | 102 | |||
| BC030387.1 | - | 5 - 6 | 391 | 102 | ||
| BC056937.1 | - | 7 - 8 | 747 | 5716144 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt2b
Genomic Map