Assay Details
| Gene Symbol: Trmt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167994.1 | NP_001161466.1 | 9 - 10 | 1052 | 67 | |
| NM_172540.2 | NP_766128.2 | 8 - 9 | 976 | 67 | ||
| GenBank mRNA | AK028999.1 | - | 8 - 9 | 932 | 67 | |
| AK049523.1 | - | 8 - 9 | 974 | 67 | ||
| AK129030.1 | - | 9 - 10 | 995 | 67 | ||
| AK137595.1 | - | 938 | 67 | |||
| BC030387.1 | - | 7 - 8 | 637 | 67 | ||
| BC056937.1 | - | 9 - 10 | 993 | 5716144 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt2b
Genomic Map