Assay Details
| Gene Symbol: Cnksr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172546.2 | NP_766134.1 | 3 - 4 | 972 | 117 | |
| GenBank mRNA | AK033015.1 | - | 3 - 4 | 973 | 117 | |
| AK149951.1 | - | 2 - 3 | 271 | 117 | ||
| AY151137.1 | - | 3 - 4 | 934 | 117 | ||
| BC024086.1 | - | 2 - 3 | 320 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnksr3
Genomic Map