Assay Details
| Gene Symbol: Serinc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172588.2 | NP_766176.1 | 2 - 3 | 317 | 79 | |
| GenBank mRNA | AB029501.1 | - | 2 - 3 | 79 | ||
| AK037699.1 | - | 2 - 3 | 317 | 79 | ||
| AK054475.1 | - | 2 - 3 | 328 | 79 | ||
| AK136799.1 | - | 2 - 3 | 365 | 79 | ||
| AK163675.1 | - | 2 - 3 | 282 | 79 | ||
| AK165334.1 | - | 2 - 3 | 324 | 79 | ||
| BC049189.1 | - | 2 - 3 | 317 | 6311617 | 79 | |
| BC062131.1 | - | 2 - 3 | 317 | 6514513 | 79 | |
More Information
Set Membership: |
|

Serinc5
Genomic Map