Assay Details
| Gene Symbol: Serinc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172588.2 | NP_766176.1 | 8 - 9 | 1112 | 65 | |
| GenBank mRNA | AB029501.1 | - | 8 - 9 | 1001 | 65 | |
| AK037699.1 | - | 8 - 9 | 1112 | 65 | ||
| AK054475.1 | - | 8 - 9 | 1123 | 65 | ||
| AK136799.1 | - | 8 - 9 | 1160 | 65 | ||
| AK165334.1 | - | 8 - 9 | 1119 | 65 | ||
| BC049189.1 | - | 8 - 9 | 1112 | 6311617 | 65 | |
| BC062131.1 | - | 8 - 9 | 1112 | 6514513 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serinc5
Genomic Map