Assay Details
| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172627.3 | NP_766215.1 | 5 - 6 | 647 | 121 | |
| GenBank mRNA | AK028188.1 | - | 5 - 6 | 506 | 121 | |
| AK081731.1 | - | 5 - 6 | 648 | 121 | ||
| BC141021.1 | - | 5 - 6 | 645 | 9055633 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pggt1b
Genomic Map