Assay Details
| Gene Symbol: Slco4c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172658.3 | NP_766246.1 | 6 - 7 | 1186 | 147 | |
| GenBank mRNA | AK049253.1 | - | 6 - 7 | 1186 | 147 | |
| AK049944.1 | - | 1151 | 147 | |||
| AY654588.1 | - | 6 - 7 | 1186 | 147 | ||
| BC119022.1 | - | 6 - 7 | 1145 | 8733771 | 147 | |
More Information
Set Membership: |
|

Slco4c1
Genomic Map