Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166633.2 | NP_001160105.1 | 2 - 3 | 236 | 70 | |
| NM_001290773.1 | NP_001277702.1 | 2 - 3 | 236 | 70 | ||
| NM_001290774.1 | NP_001277703.1 | 2 - 3 | 236 | 70 | ||
| NM_001290775.1 | NP_001277704.1 | 2 - 3 | 236 | 70 | ||
| NM_172670.3 | NP_766258.2 | 2 - 3 | 236 | 70 | ||
| GenBank mRNA | AK053214.1 | - | 2 - 3 | 248 | 70 | |
| AK077630.1 | - | 2 - 3 | 190 | 70 | ||
| AK081797.1 | - | 2 - 3 | 213 | 70 | ||
| AK220181.1 | - | 1 - 2 | 67 | 70 | ||
| AY742914.1 | - | 2 - 3 | 212 | 70 | ||
| AY742915.1 | - | 2 - 3 | 212 | 70 | ||
| BC058199.1 | - | 2 - 3 | 197 | 70 | ||
| BC096655.1 | - | 1 - 2 | 163 | 5057489 | 70 | |
| BC116715.1 | - | 1 - 2 | 165 | 40093557 | 70 | |
| CF170217.1 | - | 2 - 3 | 215 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK077630.1,NM_001290774.1,NM_001290775.1 transcripts.
Set Membership: |
|

Gyltl1b
Genomic Map