Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290774.1 | NP_001277703.1 | 3 - 4 | 330 | 77 | |
| GenBank mRNA | AK077630.1 | - | 3 - 4 | 284 | 77 | |
More Information
Set Membership: |
|

Gyltl1b
Genomic Map