Assay Details
| Gene Symbol: Fam214b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253353.1 | NP_001240282.1 | 4 - 5 | 1510 | 63 | |
| NM_001253354.1 | NP_001240283.1 | 5 - 6 | 1287 | 63 | ||
| NM_172691.3 | NP_766279.1 | 4 - 5 | 1291 | 63 | ||
| GenBank mRNA | AK045811.1 | - | 4 - 5 | 1275 | 63 | |
| AK154748.1 | - | 3 - 4 | 1626 | 63 | ||
| AK161380.1 | - | 4 - 5 | 1218 | 63 | ||
| BC046771.1 | - | 5 - 6 | 1277 | 5703628 | 63 | |
| BC071191.1 | - | 2 - 3 | 979 | 6467750 | 63 | |
More Information
Set Membership: |
|

Fam214b
Genomic Map