| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400020432.mm.1 |
NM_001253353.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020432.mm.1 |
NM_001253353.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020432.mm.1 |
NM_001253354.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020432.mm.1 |
NM_001253354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020432.mm.1 |
NM_172691.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020432.mm.1 |
NM_172691.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020432.mm.1 |
TC0400002517.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
NM_001253353.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
NM_001253353.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
NM_001253354.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
NM_001253354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
NM_172691.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
NM_172691.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
NR_045567.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
NR_045567.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020425.mm.1 |
TC0400002517.mm.1 |
MTA TC |